Learn More
Aviva Systems Biology Corp SYNCRIP Antibody - middle region (ARP40641_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40641_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SYNCRIP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SYNCRIP. Alternate names: GRY-RBP, NSAP1, RP1-3J17.2, dJ3J17.2, pp68, PP68, GRYRBP, HNRPQ1, hnRNP-QApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-57564