Learn More
Aviva Systems Biology Corp SYNCRIP Antibody - N-terminal region (ARP40640_T100) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40640_T100
Encompass_Preferred
This is a rabbit polyclonal antibody against SYNCRIP. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SYNCRIP. Alternate names: PP68, NSAP1, GRYRBP, HNRPQ1, GRY-RBP, hnRNP-QApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53219