Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Syndecan-1/CD138 protein (mouse) | 50 UG
SDP

Catalog No. 5000292260
Encompass_Preferred

Recombinant mouse Syndecan-1 (CD138) extracellular domain expressed in HEK293 cells and supplied with a C-terminal His tag in PBS solution. Designed for biochemical and cell-based assays involving Syndecan-1 interactions and detection.

  • Mouse origin, extracellular domain
  • Expressed in HEK293 mammalian cells
  • C-terminal His tag for detection or purification
  • Molecular weight approximately 45-60 kDa
  • Endotoxin level <1 EU/μg
  • Supplied in PBS, pH 7.4, 0.22 μm filtered
  • Store at -80°C; avoid repeated freeze-thaw cycles
  • Suitable for biochemical and cell-based assays

Catalog No. 50-002-92260 Supplier Medchemexpress LLC Supplier No. HYP7610150UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse Syndecan-1 (CD138) extracellular domain expressed in HEK293 cells and supplied with a C-terminal His tag in PBS solution. Designed for biochemical and cell-based assays involving Syndecan-1 interactions and detection.

  • Mouse origin, extracellular domain
  • Expressed in HEK293 mammalian cells
  • C-terminal His tag for detection or purification
  • Molecular weight approximately 45-60 kDa
  • Endotoxin level <1 EU/μg
  • Supplied in PBS, pH 7.4, 0.22 μm filtered
  • Store at -80°C; avoid repeated freeze-thaw cycles
  • Suitable for biochemical and cell-based assays

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.