Learn More
Sino Biological Anti-Syndecan-1/CD138 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 50641RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Mouse Syndecan-1 / SDC1 / CD138 (rM Syndecan-1 / SDC1 / CD138; Catalog#50641-M08H; NP_035649.1; Met 1-Glu 252). Syndecan-1 / SDC1 / CD138 specific IgG was purified by Mouse Syndecan-1 / SDC1 / CD138 affinity chromatography.
Catalog No. NC1392784