Learn More
Medchemexpress LLC Syndecan-4 Human 500ug

Supplier: Medchemexpress LLC HYP71927500UG
Recombinant human Syndecan-4 extracellular domain (residues E19-E145) supplied lyophilized for research use. The protein is provided in a tag-free format, formulated from PBS with stabilizing excipients, and characterized by SDS-PAGE and molecular weight analysis. Store frozen and aliquot for extended storage to preserve activity and integrity.
- Recombinant extracellular domain (E19-E145) for biochemical studies.
- Tag-free format suitable for binding assays and ELISA.
- Expressed in E. coli and purified to greater than 95% purity.
- Lyophilized formulation with trehalose for enhanced stability.
- Predicted molecular weight ~13.9 kDa; observed ~24 kDa on SDS-PAGE.
- Store at -20°C and aliquot for long-term storage at -20°C or -80°C.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.