Learn More
ABclonal Technology SYNE3 Rabbit pAb
SDP
Catalog No. 5000097760
Encompass_Preferred
Enables actin filament binding activity and cytoskeleton-nuclear membrane anchor activity. Involved in cytoskeleton organization, establishment of protein localization to membrane, and regulation of cell shape. Located in nuclear membrane. Part of meiotic nuclear membrane microtubule tethering complex. Biomarker of Huntingtons disease.
Catalog No. 50-000-97760 Supplier ABclonal Technology Supplier No. A13237100UL
Enables actin filament binding activity and cytoskeleton-nuclear membrane anchor activity. Involved in cytoskeleton organization, establishment of protein localization to membrane, and regulation of cell shape. Located in nuclear membrane. Part of meiotic nuclear membrane microtubule tethering complex. Biomarker of Huntingtons disease.