Learn More
TARGETMOL CHEMICALS INC Synephrine 100MG
SDP
Catalog No. 502875912
Encompass_Preferred
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Synephrine (Oxedrine) is sympathetic alpha-adrenergic agonist used as a vasoconstrictor in circulatory failure, asthma, nasal congestion, and glaucoma. Purity 99.63%
Catalog No. 50-287-5912 Supplier TARGETMOL CHEMICALS INC Supplier No. T1287100MG
Also available in 1 mL, 10 mg, 25 mg, 50 mg, 500 mg and bulk. Please contact Fisher for quotes. Synephrine (Oxedrine) is sympathetic alpha-adrenergic agonist used as a vasoconstrictor in circulatory failure, asthma, nasal congestion, and glaucoma. Purity 99.63%