Learn More
Medchemexpress LLC Synephrine | 94-07-5 | MFCD00002370 | 98.0% | 167.21 g·mol⁻1 | C9H13NO2 | 10 MG

Supplier: Medchemexpress LLC HYN013210MG
Synephrine is an alkaloid adrenergic agonist used as a research reagent for in vitro and in vivo studies. It acts as an α- and β-adrenergic agonist and is supplied as a high-purity analytical compound; consult the SDS and handling guidance before use.
- High purity (98.0%) analytical reagent for research use only.
- Reported activity: EC50 ~6.6 μM at β-adrenergic receptor in HEK293 cells.
- Available as solid (5 mg-5 g) and solution (10 mM in DMSO, 1 mL).
- Contains CAS number 94-07-5 and molecular weight 167.21 g·mol⁻1.
- Handling recommendations: use newly opened DMSO, aliquot stock solutions, store at -80°C (up to 6 months) or -20°C (up to 1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.