Learn More
Selleck Chemical LLC Synephrine HCl S2438-5mg
SDP
Supplier:  Selleck Chemical LLC S24385MG
Encompass_Preferred
SynephrineHCl (Oxedrine p-Synephrine) is a sympathomimetic -adrenergic receptor (AR) agonist
Catalog No. 50-311-1630