Learn More
Apexbio Technology LLC Synephrine HCl 5985-28-4 500mg
SDP
Catalog No. 5000089789
Encompass_Preferred
Synephrine HCl (CAS 5985-28-4) also known as Oxedrine or p-Synephrine is a small molecule that functions as a sympathomimetic agent by activating -adrenergic receptors Through its agonist activity at these receptors Synephrine HCl modulates adrenergic signaling pathways involved in vasoconstriction and other physiological processes It is commonly utilized in biomedical research to investigate adrenergic receptor pharmacology signal transduction mechanisms and cardiovascular responses associated with adrenergic stimulation
Catalog No. 50-000-89789 Supplier Apexbio Technology LLC Supplier No. B1370500
Synephrine HCl (CAS 5985-28-4) also known as Oxedrine or p-Synephrine is a small molecule that functions as a sympathomimetic agent by activating -adrenergic receptors Through its agonist activity at these receptors Synephrine HCl modulates adrenergic signaling pathways involved in vasoconstriction and other physiological processes It is commonly utilized in biomedical research to investigate adrenergic receptor pharmacology signal transduction mechanisms and cardiovascular responses associated with adrenergic stimulation