Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Synephrine Tartrate 98.0+%, TCI America™
SDP

Catalog No. S023325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S023325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S023325G Supplier TCI America Supplier No. S023325G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16589-24-5
Molecular Formula C22H32N2O10
Molecular Weight (g/mol) 484.502
MDL Number MFCD00151120
InChI Key KZZBAIXGUQOHKI-UHFFFAOYSA-N
Synonym 4-[1-Hydroxy-2-(methylamino)ethyl]phenol Tartrate, 1-(4-Hydroxyphenyl)-2-(methylamino)ethanol Tartrate, 4-Hydroxy-alpha-(methylaminomethyl)benzyl Alcohol Tartrate
PubChem CID 44135848
IUPAC Name 2,3-dihydroxybutanedioic acid;4-[1-hydroxy-2-(methylamino)ethyl]phenol
SMILES CNCC(C1=CC=C(C=C1)O)O.CNCC(C1=CC=C(C=C1)O)O.C(C(C(=O)O)O)(C(=O)O)O

Specifications

Melting Point 184°C
Color White
Quantity 25 g
Formula Weight 484.50
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Synephrine Tartrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.