Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Synergi 10 µm Hydro-RP 80 Å

Catalog No. 502393685 Shop All Phenomenex Products
Encompass

Synergi 10 µm Hydro-RP 80 Å LC Column 250 x 21.2 mm, AXIA™ Packed

Catalog No. 50-239-3685 Supplier Phenomenex Inc Supplier No. 00G4376P0AX
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synergi 10 µm Hydro-RP 80 Å LC Column 250 x 21.2 mm, AXIA™ Packed

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.