Learn More
Ops Diagnostics Synergy™ 2.0 Plant DNA Extraction Kit
Catalog No. NC1206062
Encompass
OPS Diagnostics’ Synergy™ DNA extraction platform features proprietary technology to isolate high quality DNA in a timely and cost-efficient manner without the use of hazardous chloroform. Building upon the same proprietary nucleic isolation chemistry found in the original Synergy™ Plant DNA Extraction Kit, the Synergy™ 2.0 Plant DNA Extraction Kit includes additional Silica Spin Columns to extract plant DNA for NGS and other downstream applications requiring the highest purity levels. Sample processing time is 45 minutes or less, with yields and purities exceeding those generated by traditional homegrown CTAB protocols and commercially-available kits.
Catalog No. NC1206062 Supplier Ops Diagnostics Supplier No. SYNP 0210002
OPS Diagnostics’ Synergy™ DNA extraction platform features proprietary technology to isolate high quality DNA in a timely and cost-efficient manner without the use of hazardous chloroform. Building upon the same proprietary nucleic isolation chemistry found in the original Synergy™ Plant DNA Extraction Kit, the Synergy™ 2.0 Plant DNA Extraction Kit includes additional Silica Spin Columns to extract plant DNA for NGS and other downstream applications requiring the highest purity levels. Sample processing time is 45 minutes or less, with yields and purities exceeding those generated by traditional homegrown CTAB protocols and commercially-available kits.