Learn More
Description
Predict vancomycin and high-level aminoglycoside resistance in Enterococcus spp. using Thermo Scientific™ Synergy Quad.
- One quadrant of the plate contains growth control factor
- One quadrant contains Vancomycin 6ug/mL
- One quadrant contains Gentamicin 500ug/mL
- One quadrant contains Streptomycin 2000μg/mL
Quadrant I: Control quadrant. Growth demonstrates viability of the test isolate. If no growth, the test is invalid.
Quadrant II, III, IIII: Growth indicates synergy cannot be predicted in combination therapy. No growth indicates synergy can be predicted.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Color | Clear agar |
| Content And Storage | 2°C to 8°C |
| Description | Synergy Quad |
| Format | 85mm Quadplate |
| Manufacturing Origin | United States |
| Product Type | Agar Plate |
| Fill Volume | 5 mL |
| Packaging Type | Flow wrapped - single layer |
| Quantity | 10/Pk. |
d
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
