Learn More
Cayman Chemical Synga oblta lndl Syngtn 10mg
SDP
Catalog No. 502919068
Encompass_Preferred
A methylated flavonol with diverse biological activities; inhibits α-glucosidase and scavenges radicals in a TEAC assay (IC50s = 36.8 and 12.3 µM, respectively); reduces the proliferation of, induces cell cycle arrest at the G2/M phase in, and decreases cyclin D1 and COX-2 levels in, Caco-2 cells at 50 µM; increases the lifespan of C. elegans at 100 µM
Catalog No. 50-291-9068 Supplier Cayman Chemical Supplier No. 3924910 MG
A methylated flavonol with diverse biological activities; inhibits α-glucosidase and scavenges radicals in a TEAC assay (IC50s = 36.8 and 12.3 µM, respectively); reduces the proliferation of, induces cell cycle arrest at the G2/M phase in, and decreases cyclin D1 and COX-2 levels in, Caco-2 cells at 50 µM; increases the lifespan of C. elegans at 100 µM