Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syngene™ Neutral Field Screen Set for use with IG3 and UG3 systems
GREENER_CHOICESDP

Catalog No. 01257233 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
Syngene Ingenius and Nugenius Systems
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-257-233 Syngene Ingenius and Nugenius Systems
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-257-233 Supplier Syngene™ Supplier No. GENFS
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Contains a blue screen and a frosted screen. Syngene™ Neutral Field Screen Set is for use with IG3 and UG3 systems.

  • Corrects for any uneven Illumination by acquiring a neutral-field reference image with the same intensity illumination as the experiment.
  • For Epi-UV lighting and transilluminators in Syngene™ Ingenius and Nugenius Systems

Specifications

Product Type Neutral Field Screen Set
For Use With (Application) IG3 and UG3 Systems
For Use With (Equipment) Syngene Ingenius and Nugenius Systems
Dimensions (L x W x H) 330 x 310 x 15 mm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.