Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Syngene™ Long Wave UV Tube
GREENER_CHOICESDP

Catalog No. 01257291 Shop All Synoptics, Inc. Products
Change view
Click to view available options
Wavelength:
365 nm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wavelength
01-257-291 365 nm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-257-291 Supplier Syngene™ Supplier No. LWT8
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Syngene™ Long Wave UV Tube for use in UV Transilluminators.

  • For use with all GX and GE model transilluminators
  • 365nm wavelength
  • Long wave

Specifications

Wavelength 365 nm
Type Longwave UV Tube
For Use With (Equipment) All GX/GE model transilluminators
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.