Learn More
Syngene™ Visible Light Converter
GREENER_CHOICESDP
Catalog No. p-8011046 Shop All Synoptics, Inc. Products
Change view
Click to view available options
For Use With (Equipment):
G:BOX Systems
IG/UG and DG Systems
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment)
01-257-231 IG/UG and DG Systems
01-257-271 G:BOX Systems
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-257-231 Supplier Syngene™ Supplier No. GECONVERT5
Add to Cart
Add to Cart
Use the Visible Light Converter in conjunction with your UV transilluminator. Screen provides a white surface for visualising visible protein gels.
  • Use the Visible light converter in conjunction with a UV transilluminator provides a white surface for visualising visible protein gels.
Specifications
Intensity Fixed
Type Converter Plate
Description UV (White)
Wavelength 302 nm
For Use With (Equipment) IG/UG and DG Systems
WARNING: Cancer - www.P65Warnings.ca.gov