Learn More
Aviva Systems Biology Corp SYNGR2 Antibody - N-terminal region (ARP46485_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46485_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SYNGR2. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SYNGR2. Alternate names: MGC102914ApplicationWB. Species reactivity: Cow, Human, Mouse, Pig, Rabbit, Rat, Zebrafish
Catalog No. 50-014-59340