Learn More
Aviva Systems Biology Corp SYNJ1 Antibody - middle region (ARP41223_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41223_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SYNJ1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human SYNJ1. Alternate names: INPP5GApplicationWB. Species reactivity: Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57768