Learn More
Aviva Systems Biology Corp SYNJ1 Antibody - N-terminal region (ARP40498_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP40498_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against SYNJ1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human SYNJ1. Alternate names: INPP5GApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-57488