Learn More
Sigma Aldrich Fine Chemicals Biosciences SynLectroTM Boron doped Di1EA
Catalog No. 502909106
Encompass
SynLectro(TM) Boron-doped Diamond Electrode is a 3x20x60mm electrode designed for use with the SynLectro(TM) Electrolysis Platform for synthetic electrochemistry.SynLectro(TM) Electrolysis PlatformThe SynLectro(TM) Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component designed with consistency reproducibility and safety in mind.
Catalog No. 50-290-9106 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. ESYNTH0201EA
SynLectro(TM) Boron-doped Diamond Electrode is a 3x20x60mm electrode designed for use with the SynLectro(TM) Electrolysis Platform for synthetic electrochemistry.SynLectro(TM) Electrolysis PlatformThe SynLectro(TM) Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component designed with consistency reproducibility and safety in mind.