Learn More
Sigma Aldrich Fine Chemicals Biosciences SynLectroTM Glassy Carbon1EA
Catalog No. 502904800
Encompass
SynLectro(TM) Glassy Carbon Electrode is a 3x20x60 mm electrode designed for use with the SynLectro(TM) Electrolysis Platform for synthetic electrochemistry.SynLectro(TM) Electrolysis PlatformThe SynLectro(TM) Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component designed with consistency reproducibility and safety in mind.
Catalog No. 50-290-4800 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. ESYNTH0191EA
SynLectro(TM) Glassy Carbon Electrode is a 3x20x60 mm electrode designed for use with the SynLectro(TM) Electrolysis Platform for synthetic electrochemistry.SynLectro(TM) Electrolysis PlatformThe SynLectro(TM) Electrolysis Platform is a portfolio of labware products and electrodes designed to simplify and standardize preparative scale electrosynthesis. Learn more about each component designed with consistency reproducibility and safety in mind.