Learn More
Med Vet International Zoetis, Synotic Otic Soln, 8mL, 12/Case
Catalog No. 502828925
Encompass_Preferred
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Contains fluocinolone acetonide 0.01% and dimethyl sulfoxide 60%. Indicated for the relief of pruritus and inflammation associated with acute and chronic otitis in dogs.
Catalog No. 50-282-8925 Supplier Med Vet International Supplier No. RXSYN8
VET LICENSE NEEDS TO BE PROVIDED IN 3-4 BUSINESS DAYS OR ORDER WILL BE CANCELLED
Contains fluocinolone acetonide 0.01% and dimethyl sulfoxide 60%. Indicated for the relief of pruritus and inflammation associated with acute and chronic otitis in dogs.