Learn More
Sklar Instruments Synovectomy Rongeur, Sklar, Strong Curve, 2mm, Stainless Steel. EA/1.
SDP
Catalog No. 501438509 Shop All Sklar Instruments Products
Encompass_Preferred
Synovectomy Rongeur, Sklar, Strong Curve, 2mm, Stainless Steel. Sklar's Synovectomy Rongeur is an orthopedic instrument used in synovectomy procedures, primarily to treat rheumatoid arthritis. This product is a non-ratcheted, finger ring instrument that uses a small, curved, scooping tip to remove synovium that has become inflamed or thickened and thus not providing the necessary lubrication to the small joints in hands or feet. This product has strongly curved jaws with 2mm tips. The overall length is 5 inches.
This item is not returnable. View return policy
Catalog No. 50-143-8509 Supplier Sklar Instruments Supplier No. 403213
Synovectomy Rongeur, Sklar, Strong Curve, 2mm, Stainless Steel. Sklar's Synovectomy Rongeur is an orthopedic instrument used in synovectomy procedures, primarily to treat rheumatoid arthritis. This product is a non-ratcheted, finger ring instrument that uses a small, curved, scooping tip to remove synovium that has become inflamed or thickened and thus not providing the necessary lubrication to the small joints in hands or feet. This product has strongly curved jaws with 2mm tips. The overall length is 5 inches.