Learn More
Med Vet International Synovex S, (Progesterone And Estradiol Benzoate), Steer Implant, 100 Count
Catalog No. 502827789
Encompass_Preferred
Steer implants for increased rate of weight gain and improved feed efficiency. One dose of 8 pellets contains 200 mg progesterone and 20 mg estradiol benzoate per implantation.
Catalog No. 50-282-7789 Supplier Med Vet International Supplier No. PHVSYNOVEXS
Steer implants for increased rate of weight gain and improved feed efficiency. One dose of 8 pellets contains 200 mg progesterone and 20 mg estradiol benzoate per implantation.