Learn More
Sciencell Research Laboratories Synoviocyte Medium
Catalog No. NC1621843
Encompass
SM consists of 500 ml of basal medium, 10 ml of fetal bovine serum (FBS, Cat. No. 0010), 5 ml of synoviocyte growth supplement (SGS, Cat. No. 4752), and 5 ml of penicillin/streptomycin solution (P/S, Cat. No. 0503).
Catalog No. NC1621843 Supplier Sciencell Research Laboratories Supplier No. 4701
SM consists of 500 ml of basal medium, 10 ml of fetal bovine serum (FBS, Cat. No. 0010), 5 ml of synoviocyte growth supplement (SGS, Cat. No. 4752), and 5 ml of penicillin/streptomycin solution (P/S, Cat. No. 0503).