Learn More
Crescent Chemical Co Inc SYNPERONIC F108 PRACT.
SDP
Catalog No. 50248176
Encompass_Preferred
Synperonic F108 pract. Polypropylene glycol (Mr ca. 3 250): poly(ethylene glycol) ca. 1:4. MP 55 - 60 C. Strongly hydrophilic (HLB ca. 27). More than 10 % soluble in water. Size - 1KG Storage Conditions - +15 C TO +30 C Catalog Number - 35726.02
Catalog No. 50-248-176 Supplier Crescent Chemical Co Inc Supplier No. SE35726.02
Synperonic F108 pract. Polypropylene glycol (Mr ca. 3 250): poly(ethylene glycol) ca. 1:4. MP 55 - 60 C. Strongly hydrophilic (HLB ca. 27). More than 10 % soluble in water. Size - 1KG Storage Conditions - +15 C TO +30 C Catalog Number - 35726.02