Learn More
Crescent Chemical Co Inc Synperonic F108, 100g
SDP
Catalog No. 50248175
Encompass_Preferred
Synperonic F108 pract. Polypropylene glycol (Mr ca. 3 250): poly(ethylene glycol) ca. 1:4. MP 55 - 60 C. Strongly hydrophilic (HLB ca. 27). More than 10 % soluble in water. Storage Conditions - +15 C TO +30 C
Catalog No. 50-248-175 Supplier Crescent Chemical Co Inc Supplier No. SE35726.01
Synperonic F108 pract. Polypropylene glycol (Mr ca. 3 250): poly(ethylene glycol) ca. 1:4. MP 55 - 60 C. Strongly hydrophilic (HLB ca. 27). More than 10 % soluble in water. Storage Conditions - +15 C TO +30 C