Learn More
Crescent Chemical Co Inc SYNPERONIC F68 PRACT.
SDP
Catalog No. 50248174
Encompass_Preferred
Synperonic F68 pract. Polyol detergent, block-copolymer. Pluronic™ PE 6800. Poloxamer 188. Poly(propylene glycol) (Mr ca. 1 750): polyethylene glycol ca. 1:4.MP ca. 55 C.Strongly hydrophilic detergent (HLB approx. 29). More than 10 % soluble in water, forms foa
Catalog No. 50-248-174 Supplier Crescent Chemical Co Inc Supplier No. SE35724.02
Synperonic F68 pract. Polyol detergent, block-copolymer. Pluronic™ PE 6800. Poloxamer 188. Poly(propylene glycol) (Mr ca. 1 750): polyethylene glycol ca. 1:4.MP ca. 55 C.Strongly hydrophilic detergent (HLB approx. 29). More than 10 % soluble in water, forms foa