Learn More
Crescent Chemical Co Inc Synperonic F68, 100g
SDP
Catalog No. 50248173
Encompass_Preferred
Polyol detergent, block-copolymer. Pluronic™ PE 6800. Poloxamer 188. Poly(propylene glycol) (Mr ca. 1 750): polyethylene glycol ca. 1:4.MP ca. 55 °C.Strongly hydrophilic detergent (HLB approx. 29). More than 10 % soluble in water, forms foams moderately. Used in cell culture to protect microorganisms, animal and plant cells against mechanical damage. Storage Conditions - +15 °C TO +30 °C
Catalog No. 50-248-173 Supplier Crescent Chemical Co Inc Supplier No. SE35724.01
Polyol detergent, block-copolymer. Pluronic™ PE 6800. Poloxamer 188. Poly(propylene glycol) (Mr ca. 1 750): polyethylene glycol ca. 1:4.MP ca. 55 °C.Strongly hydrophilic detergent (HLB approx. 29). More than 10 % soluble in water, forms foams moderately. Used in cell culture to protect microorganisms, animal and plant cells against mechanical damage. Storage Conditions - +15 °C TO +30 °C