Learn More
ABclonal Technology SYNPO Rabbit pAb
SDP
Supplier:  ABclonal Technology A12049500UL
Encompass_Preferred
Synaptopodin is an actin-associated protein that may play a role in actin-based cell shape and motility. The name synaptopodin derives from the proteins associations with postsynaptic densities and dendritic spines and with renal podocytes (Mundel et al., 1997 [PubMed 9314539]).
Catalog No. 50-000-99049