Learn More
Electron Microscopy Sciences Synaptek DOT Grid with 1.5 mm hole, Beryllium-Copper
SDP
Catalog No. 501926632
Encompass_Preferred
Synaptek unflexible grids are made of a special Beryllium-Copper alloy and very reliable under the electron beam. Offer extreme stability for coating with support film. 4 mil thick (100um), 3.05 mm diameter. Contamination free and reusable after cleaning.
I.D.: 1.5 mm hole
Material: Beryllium-Copper
Catalog No. 50-192-6632 Supplier Electron Microscopy Sciences Supplier No. S1500DOT
Synaptek unflexible grids are made of a special Beryllium-Copper alloy and very reliable under the electron beam. Offer extreme stability for coating with support film. 4 mil thick (100um), 3.05 mm diameter. Contamination free and reusable after cleaning.
I.D.: 1.5 mm hole
Material: Beryllium-Copper