Learn More
Phenomenex Inc Phenomenex Synergi Column, Hydro-RP, 10 μm, 250 x 4.6 mm
Catalog No. NC3713216 Shop All Phenomenex Products
Encompass
Synergi Hydro-RP HPLC column optimized for retention and separation of polar and weakly hydrophobic compounds, with compatibility for highly aqueous mobile phases.
  • Hydro-RP stationary phase for improved polar analyte retention.
  • 10 μm particle size for robust, high-capacity separations.
  • 80 Å pore size suitable for small to medium molecules.
  • 250 x 4.6 mm dimensions for analytical-scale runs.
  • Reversed-phase HPLC mode, compatible with 100% aqueous solvents.
  • Approximately 19% carbon load for balanced retention and efficiency.
Catalog No. NC3713216 Supplier Phenomenex Inc Supplier No. 00G4376EO
Synergi Hydro-RP HPLC column optimized for retention and separation of polar and weakly hydrophobic compounds, with compatibility for highly aqueous mobile phases.
  • Hydro-RP stationary phase for improved polar analyte retention.
  • 10 μm particle size for robust, high-capacity separations.
  • 80 Å pore size suitable for small to medium molecules.
  • 250 x 4.6 mm dimensions for analytical-scale runs.
  • Reversed-phase HPLC mode, compatible with 100% aqueous solvents.
  • Approximately 19% carbon load for balanced retention and efficiency.