Learn More
Medchemexpress LLC Synstatin (92-119) | 1259384-47-8 | 99.4% | 3032.27 | 10 MG
SDP
Catalog No. 5000399943
Encompass_Preferred
Synstatin (92-119) is a synthetic peptide that acts as an inhibitor of αvβ3/αvβ5 integrins and IGF1R, exhibiting anti-angiogenic, anti-proliferative, antioxidant, and anti-tumor activities. It competitively blocks the capture of αvβ3/αvβ5 integrins and IGF1R by syndecan-1, disrupting the formation of the syndecan-1 : integrin : IGF1R ternary complex and inhibiting integrin activation. This compound is delivered as a white to off-white solid, readily soluble in DMSO and H2O.
  • Inhibitor of αvβ3/αvβ5 integrins and IGF1R
  • Exhibits anti-angiogenic, anti-proliferative, antioxidant, and anti-tumor effects
  • Disrupts syndecan-1:integrin:IGF1R ternary complex formation
  • Inhibits integrin activation and talin-mediated signaling pathways
  • Blocks VEGF-induced angiogenesis
  • White to off-white solid appearance
  • Soluble in DMSO and H2O
Catalog No. 50-003-99943 Supplier Medchemexpress LLC Supplier No. HYP513310MG
Synstatin (92-119) is a synthetic peptide that acts as an inhibitor of αvβ3/αvβ5 integrins and IGF1R, exhibiting anti-angiogenic, anti-proliferative, antioxidant, and anti-tumor activities. It competitively blocks the capture of αvβ3/αvβ5 integrins and IGF1R by syndecan-1, disrupting the formation of the syndecan-1 : integrin : IGF1R ternary complex and inhibiting integrin activation. This compound is delivered as a white to off-white solid, readily soluble in DMSO and H2O.
  • Inhibitor of αvβ3/αvβ5 integrins and IGF1R
  • Exhibits anti-angiogenic, anti-proliferative, antioxidant, and anti-tumor effects
  • Disrupts syndecan-1:integrin:IGF1R ternary complex formation
  • Inhibits integrin activation and talin-mediated signaling pathways
  • Blocks VEGF-induced angiogenesis
  • White to off-white solid appearance
  • Soluble in DMSO and H2O