Learn More
Medchemexpress LLC Synstatin (92-119) | 1259384-47-8 | 99.4% | 3032.27 | 5 MG
SDP
Catalog No. 5000398482
Encompass_Preferred
Synstatin (92-119) is an inhibitor of αvβ3/αvβ5 integrins and IGF1R, exhibiting anti-angiogenic, anti-proliferative, antioxidant, and anti-tumor activities. It competitively blocks the capture of αvβ3/αvβ5 integrins and IGF1R by syndecan-1, disrupting the formation of the syndecan-1: integrin: IGF1R ternary complex. It also inhibits integrin activation, talin-mediated signaling, and VEGF-induced angiogenesis, making it suitable for cancer and hepatocellular carcinoma research.
  • Directly inhibits assembly of Sdc1:αvβ3 integrin:IGF1R ternary complex
  • Binds to αvβ3 integrin, αvβ5 integrin, and IGF1R on activated vascular endothelial cells
  • Inhibits αvβ3-dependent adhesion and migration of tumor cells
  • Inhibits VEGF- and αvβ3-dependent angiogenesis
  • Reduces hepatocellular carcinoma progression in thioacetamide-induced rats
Catalog No. 50-003-98482 Supplier Medchemexpress LLC Supplier No. HYP51335MG
Synstatin (92-119) is an inhibitor of αvβ3/αvβ5 integrins and IGF1R, exhibiting anti-angiogenic, anti-proliferative, antioxidant, and anti-tumor activities. It competitively blocks the capture of αvβ3/αvβ5 integrins and IGF1R by syndecan-1, disrupting the formation of the syndecan-1: integrin: IGF1R ternary complex. It also inhibits integrin activation, talin-mediated signaling, and VEGF-induced angiogenesis, making it suitable for cancer and hepatocellular carcinoma research.
  • Directly inhibits assembly of Sdc1:αvβ3 integrin:IGF1R ternary complex
  • Binds to αvβ3 integrin, αvβ5 integrin, and IGF1R on activated vascular endothelial cells
  • Inhibits αvβ3-dependent adhesion and migration of tumor cells
  • Inhibits VEGF- and αvβ3-dependent angiogenesis
  • Reduces hepatocellular carcinoma progression in thioacetamide-induced rats