Learn More
Enzo Life Sciences Syntaxin 13 monoclonal antibody (15G2) (100µg)
SDP
Supplier:  Enzo Life Sciences ADIVAMSV026E
Encompass_Preferred
Clone: 15G2. Host: Mouse. Isotype: IgG. Immunogen: Recombinant cytoplasmic domain of rat syntaxin 13. Source: Purified from ascites. Species reactivity: Human, Mouse, Rat Hamster, Dog. Applications: IHC (PS), WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7703