Learn More
Enzo Life Sciences Syntaxin 2 polyclonal antibody (100µg)
SDP
Supplier:  Enzo Life Sciences ADIVAPSV065E
Encompass_Preferred
Host: Rabbit. Immunogen: Synthetic peptide corresponding to the sequence near the N-terminus of rat syntaxin 2. Source: Purified from rabbit serum. Species reactivity: Human, Mouse, Rat, Xenopus, Sheep, Rabbit, Porcine, Monkey, Hamster, Drosophila, Dog, Chicken, Bovine. Applications: IHC (PS), IP, WB. Purity: Peptide immunoaffinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7714