Learn More
Enzo Life Sciences Syntaxin 6 monoclonal antibody (3D10) (200µg)
SDP
Supplier:  Enzo Life Sciences ADIVAMSV025F
Encompass_Preferred
Clone: 3D10. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant rat Syntaxin 6. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat Hamster. Applications: IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7702