Learn More
Enzo Life Sciences Syntaxin 6 monoclonal antibody (3D10) (50µg)
SDP
Supplier:  Enzo Life Sciences ADIVAMSV025D
Encompass_Preferred
Clone: 3D10. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant rat Syntaxin 6. UniProt ID: Q63635. Source: Purified from mouse ascites. Species reactivity: Human, Mouse, Rat Hamster. Applications: IP, WB. Recommended Dilutions: Immunoprecipitation (12µg/ml) Western Blot (1:1,000, ECL) Suggested dilutions/conditions may not be available for all applications. Optimal conditions must be determined individually for each application. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide. Handling: Avoid freeze/thaw cycles.
Catalog No. 50-200-7701