Learn More
ProSci Syntenin 2 Peptide

Description
Specifications
Specifications
| Gene ID (Entrez) | 27111 |
| Species | Human |
| Accession Number | NP_056500, NP_536737 |
| Common Name | syndecan binding protein (syntenin) 2 |
| Content And Storage | Store at -20°C. When reconstituted minimize freezing and thawing. |
| Format | Lyophilized |
| Formulation | Syntenin 2 peptide is supplied as 100 μg of dried peptide. When the peptide is reconstituted in 200 μL water, the concentration will be 0.5 mg/mL |
| For Use With (Application) | Blocking Assay |
| Gene Symbol | SDCBP2 |
| Quantity | 0.1 mg |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.