Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Synthegel™ Spheroid Matrix Kit, 10 mL

Catalog No. 02707162 Shop All Corning Products
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-707-162 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-707-162 Supplier Corning™ Supplier No. 354789
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Research tool for high content 3D biologically formed spheroid culture with more accurate in vivo predictions. The matrix has a stronger gel and optimal cytocompatibility for spheroid culture. The kit consists of a Spheroid matrix peptide nanofiber solution and a Synthegel X-Link solution.

  • Matrix rigidity is tunable by altering the peptide concentration
  • Purified synthetic peptide in neutral pH
  • Cells are encapsulated in physiological mimicking conditions
  • Fast hydrogel formation (5 to 30 minutes)
  • Sol-gel transfer reversible by shearing force
  • Works with standard cell culture methodologies, growth factors, and media
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO
Color Clear
Content And Storage Store at 2 to 8°C
Culture Environment Ambient Temperature
Description Synthegel™ Spheroid Matrix Kit
Culture Type 3D Cell Culture
Form Liquid
Product Type Extracellular Matrix
Sterility Sterile-filtered
Format Kit
For Use With (Application) Culture of cancer spheroids in a synthetic peptide hydrogel
pH Neutral
Product Line Hydrogel
Quantity 10 mL
Shipping Condition Ambient Temperature
Source Synthetic Peptide Matrix
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.