Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Corning™ Synthegel™ 3D hiPSC Suspension Matrix Kit, 16 mL

Catalog No. 02707163 Shop All Corning Products
Change view
Click to view available options
Quantity:
16 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-707-163 16 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-707-163 Supplier Corning™ Supplier No. 354791
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Powerful bio-tool for large-scale manufacturing of physiological 3D hiPSC spheroids in a lab setting. After harvesting, spheroids can be used directly for various downstream tasks such as drug screening, bioprinting for tissue engineering, and somatic cell differentiation.

  • Kit consists of a vial of Synthegel hiPSC suspension matrix, a nanofiber solution for 3D suspension culture and a vial of Synthegel X-Link solution
  • Requires a vial of lyophilized Synthegel 3D hiPSC Grow Mix
  • Matrix rigidity is tunable by altering the peptide concentration
  • Purified synthetic peptide in neutral pH
  • Cells are encapsulated in physiological mimicking conditions
  • Fast hydrogel formation (5 to 30 minutes)
  • Sol-gel transfer reversible by shearing force
  • Works with standard cell culture methodologies, growth factors and media
  • Allows growth and passaging of hiPSCs in 3D format. The hiPSC Grow Mix supplement provides a complete environment for hiPSC 3D culture
  • Allows 3D culture of hiPSC in suspension
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO
Color Red
Content And Storage Store at 2 to 8°C
Culture Environment Ambient Temperature
Description Synthegel™ 3D hiPSC Suspension Matrix Kit
Culture Type 3D Cell Culture
Form Liquid
Product Type Extracellular Matrix
Sterility Sterile-filtered
Format Kit
For Use With (Application) 3D suspension culture of hiPSC in a synthetic peptide hydrogel
pH Neutral
Product Line Hydrogel
Quantity 16 mL
Shipping Condition Ambient Temperature
Source Synthetic Peptide Matrix
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.