Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Innovating Science™ Synthesis of a Coordination Complex
SCI_ED

Catalog No. S02004 Shop All Aldon Products
Change view
Click to view available options
Grade:
9 to Undergraduate
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S02004 9 to Undergraduate
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
WARNING: This product can expose you to chemicals including Methanol, Methyl Isobutyl Ketone, and strong inorganic mists containing Sulfuric Acid, which are known to the state of California to cause cancer and birth defects or reproductive harm. For more information go to www.P65Warnings.ca.gov
Catalog No. S02004 Supplier Innovating Science™ Supplier No. IS2535
Add to Cart
Add to Cart
WARNING: This product can expose you to chemicals including Methanol, Methyl Isobutyl Ketone, and strong inorganic mists containing Sulfuric Acid, which are known to the state of California to cause cancer and birth defects or reproductive harm. For more information go to www.P65Warnings.ca.gov

Synthesize and purify a lime-green coordination complex called potassium ferrioxalate trihydrate in this multi-step synthesis.

Coordination complexes are brightly colored compounds consisting of a central metal atom and ligand molecules, atoms, or ions. Synthesize and purify a lime-green coordination complex called potassium ferrioxalate trihydrate in this multi-step synthesis and optionally analyze the complex via UV-Visible spectrophotometry.

This kit contains enough materials for 15 groups, a Teacher's Manual, and Student Study Guide copymaster.

Specifications

Grade 9 to Undergraduate
Class Size 15 Lab Groups
Product Type Synthesis and Gravimetric Analysis of Cobalt Oxalate Hydrate Chemistry Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.