Learn More
Electron Microscopy Sciences Synthesized Graphene Powder
SDP
Catalog No. 5034224
Encompass_Preferred
Graphene is the ideal support film for electron microscopy. It provides an invisible, crystalline background that enables the unrivaled TEM characterization of organic and inorganic nanomaterials. Graphene is atomically thin, chemically inert, consists of light atoms, and possesses a highly ordered structure. Graphene is electrically and thermally conductive, and is the strongest material ever measured.
We offer ultrahigh-quality graphene that is produced through the substrate-free gas-phase method which results in a highly ordered structure that is composed of 99% carbon by mass (1% hydrogen).
Catalog No. 50-342-24 Supplier Electron Microscopy Sciences Supplier No. GF1205
Graphene is the ideal support film for electron microscopy. It provides an invisible, crystalline background that enables the unrivaled TEM characterization of organic and inorganic nanomaterials. Graphene is atomically thin, chemically inert, consists of light atoms, and possesses a highly ordered structure. Graphene is electrically and thermally conductive, and is the strongest material ever measured.
We offer ultrahigh-quality graphene that is produced through the substrate-free gas-phase method which results in a highly ordered structure that is composed of 99% carbon by mass (1% hydrogen).