Learn More
Cayman Chemical synthetc CnabINId ScrenI 50uL
SDP
Catalog No. 502895314
Encompass_Preferred
An LMW-PTP inhibitor (Ki 1.2 nM) greater than 8000-fold selective for LMW-PTP over 24 phosphatases at 10 UM
Catalog No. 50-289-5314 Supplier Cayman Chemical Supplier No. 900289150 UL
Add to Cart
Add to Cart
Request a Quote
An LMW-PTP inhibitor (Ki 1.2 nM) greater than 8000-fold selective for LMW-PTP over 24 phosphatases at 10 UM