Learn More
Cayman Chemical SynthetIc AlIzarIn 100g
SDP
Catalog No. 502952622
Encompass_Preferred
Inhibitor of P450 isoforms CYP1A1, CYP1A2, and CYP1B1; antioxidant activity in rats; reduces hepatic content of reactive substances and serum aminotransferases in poisoned rats; a small molecule red dye
Catalog No. 50-295-2622 Supplier Cayman Chemical Supplier No. 11676100 G
Inhibitor of P450 isoforms CYP1A1, CYP1A2, and CYP1B1; antioxidant activity in rats; reduces hepatic content of reactive substances and serum aminotransferases in poisoned rats; a small molecule red dye