Learn More
Cayman Chemical SynthetIc AlysIn 10mg
SDP
Catalog No. 502991860
Encompass_Preferred
An isothiocyanate with diverse biological activities; active against a variety of bacteria; directly inhibits PAH-induced CYP1A1 and CYP1A2 activity in MCF-7 cells at 0.5-2.5 µM; inhibits proliferation of HCT116 colon cancer cells (IC50 = 50s = 168 and 20 µM, respectively)
Catalog No. 50-299-1860 Supplier Cayman Chemical Supplier No. 3151310 MG
An isothiocyanate with diverse biological activities; active against a variety of bacteria; directly inhibits PAH-induced CYP1A1 and CYP1A2 activity in MCF-7 cells at 0.5-2.5 µM; inhibits proliferation of HCT116 colon cancer cells (IC50 = 50s = 168 and 20 µM, respectively)