Learn More
Cayman Chemical SynthetIc ArbutIn-d4 1mg
SDP
Supplier:  Cayman Chemical 332891 MG
Encompass_Preferred
An internal standard for the quantification of arbutin by GC- or LC-MS
Catalog No. 50-295-1012