Learn More
Cayman Chemical Artemitin, Purity: ≥98%, CAS: 479-90-3, 5 mg
SDP
Catalog No. 502943648
Encompass_Preferred
A flavonoid with diverse biological activities; scavenges peroxyl radicals in a cell-free assay; inhibits 5-LO (IC50 = 54.6 µM); active against P. falciparum (IC50 = 26 µM); cytotoxic to HL-60 leukemia cells (IC50 = 30.98 µM); decreases mean arterial blood pressure in normotensive rats, as well as reduces angiotensin I-induced, but not angiotensin II-induced, increases in mean arterial blood pressure in rats, at 0.75 mg/kg
Catalog No. 50-294-3648 Supplier Cayman Chemical Supplier No. 402695 MG
A flavonoid with diverse biological activities; scavenges peroxyl radicals in a cell-free assay; inhibits 5-LO (IC50 = 54.6 µM); active against P. falciparum (IC50 = 26 µM); cytotoxic to HL-60 leukemia cells (IC50 = 30.98 µM); decreases mean arterial blood pressure in normotensive rats, as well as reduces angiotensin I-induced, but not angiotensin II-induced, increases in mean arterial blood pressure in rats, at 0.75 mg/kg